Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608407 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPYSL4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DPYSL4 - dihydropyrimidinase like 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STK32C - serine/threonine kinase 32C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318878.1 | 1719 | UTR 3 | NP_001305807.1 | |||
NM_001318879.1 | 1719 | UTR 3 | NP_001305808.1 | |||
NM_001318880.1 | 1719 | Intron | NP_001305809.1 | |||
NM_001318881.1 | 1719 | Intron | NP_001305810.1 | |||
NM_001318882.1 | 1719 | Intron | NP_001305811.1 | |||
NM_001318883.1 | 1719 | Intron | NP_001305812.1 | |||
NM_173575.3 | 1719 | UTR 3 | NP_775846.2 | |||
XM_011539688.1 | 1719 | UTR 3 | XP_011537990.1 | |||
XM_011539690.2 | 1719 | UTR 3 | XP_011537992.1 | |||
XM_011539693.2 | 1719 | UTR 3 | XP_011537995.1 | |||
XM_011539694.1 | 1719 | UTR 3 | XP_011537996.1 | |||
XM_011539695.1 | 1719 | UTR 3 | XP_011537997.1 | |||
XM_011539696.1 | 1719 | UTR 3 | XP_011537998.1 | |||
XM_017016100.1 | 1719 | Intron | XP_016871589.1 | |||
XM_017016101.1 | 1719 | Intron | XP_016871590.1 |
Set Membership: |
HapMap |