Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606417 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR11 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
WDR11 - WD repeat domain 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018117.11 | Intron | NP_060587.8 | ||||
XM_005269962.2 | Intron | XP_005270019.1 | ||||
XM_005269963.2 | Intron | XP_005270020.1 | ||||
XM_005269964.2 | Intron | XP_005270021.1 | ||||
XM_017016397.1 | Intron | XP_016871886.1 | ||||
XM_017016398.1 | Intron | XP_016871887.1 | ||||
XM_017016399.1 | Intron | XP_016871888.1 | ||||
XM_017016400.1 | Intron | XP_016871889.1 |
WDR11-AS1 - WDR11 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |