Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 164920 | |||||||||||||||||||||||||||||
Literature Links: |
KIT PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
KIT - KIT proto-oncogene receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000222.2 | Intron | NP_000213.1 | ||||
NM_001093772.1 | Intron | NP_001087241.1 | ||||
XM_005265740.1 | Intron | XP_005265797.1 | ||||
XM_005265741.1 | Intron | XP_005265798.1 | ||||
XM_005265742.2 | Intron | XP_005265799.1 | ||||
XM_017008178.1 | Intron | XP_016863667.1 | ||||
XM_017008179.1 | Intron | XP_016863668.1 | ||||
XM_017008180.1 | Intron | XP_016863669.1 |
Set Membership: |
HapMap |