Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 606741 | ||||||||||||||||||||||||||
Literature Links: |
ZNF181 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ZNF181 - zinc finger protein 181 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029997.3 | 598 | Missense Mutation | CAT,TAT | H,Y 123 | NP_001025168.2 | |
NM_001145665.1 | 598 | Missense Mutation | CAT,TAT | H,Y 122 | NP_001139137.1 | |
XM_005258849.2 | 598 | Missense Mutation | CAT,TAT | H,Y 123 | XP_005258906.1 | |
XM_005258850.2 | 598 | Missense Mutation | CAT,TAT | H,Y 122 | XP_005258907.1 | |
XM_005258851.3 | 598 | Missense Mutation | CAT,TAT | H,Y 119 | XP_005258908.1 | |
XM_006723183.3 | 598 | Missense Mutation | CAT,TAT | H,Y 123 | XP_006723246.1 | |
XM_006723184.3 | 598 | Missense Mutation | CAT,TAT | H,Y 118 | XP_006723247.1 | |
XM_017026739.1 | 598 | Missense Mutation | CAT,TAT | H,Y 122 | XP_016882228.1 | |
XM_017026740.1 | 598 | Missense Mutation | CAT,TAT | H,Y 137 | XP_016882229.1 |