Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606351 | ||||||||||||||||||||
Literature Links: |
ESPN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ESPN - espin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031475.2 | Intron | NP_113663.2 | ||||
XM_005263501.2 | Intron | XP_005263558.1 | ||||
XM_011542231.1 | Intron | XP_011540533.1 | ||||
XM_011542232.1 | Intron | XP_011540534.1 | ||||
XM_011542233.2 | Intron | XP_011540535.1 | ||||
XM_011542236.2 | Intron | XP_011540538.1 | ||||
XM_011542237.1 | Intron | XP_011540539.1 | ||||
XM_011542238.2 | Intron | XP_011540540.1 | ||||
XM_017002433.1 | Intron | XP_016857922.1 | ||||
XM_017002434.1 | Intron | XP_016857923.1 |