Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTGTCCATTTCAGCCTGACTGTC[C/T]GTCTTCATGACACTCTGTGATGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602944 | ||||||||||||||||||||
Literature Links: |
E2F6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
E2F6 - E2F transcription factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278275.1 | 1563 | UTR 3 | NP_001265204.1 | |||
NM_001278276.1 | 1563 | UTR 3 | NP_001265205.1 | |||
NM_001278277.1 | 1563 | UTR 3 | NP_001265206.1 | |||
NM_001278278.1 | 1563 | UTR 3 | NP_001265207.1 | |||
NM_198256.3 | 1563 | UTR 3 | NP_937987.2 | |||
NM_212540.2 | 1563 | UTR 3 | NP_997705.1 |