Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 165070 | ||||||||||||||||||||
Literature Links: |
FLT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FLT1 - fms related tyrosine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159920.1 | 5570 | Intron | NP_001153392.1 | |||
NM_001160030.1 | 5570 | Intron | NP_001153502.1 | |||
NM_001160031.1 | 5570 | Intron | NP_001153503.1 | |||
NM_002019.4 | 5570 | UTR 3 | NP_002010.2 | |||
XM_011535014.1 | 5570 | Intron | XP_011533316.1 | |||
XM_017020485.1 | 5570 | UTR 3 | XP_016875974.1 |
PAN3 - PAN3 poly(A) specific ribonuclease subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |