Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606122 | ||||||||||||||||||||||||||||||||
Literature Links: |
TNFRSF19 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
TNFRSF19 - TNF receptor superfamily member 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204458.1 | Intron | NP_001191387.1 | ||||
NM_001204459.1 | Intron | NP_001191388.1 | ||||
NM_018647.3 | Intron | NP_061117.2 | ||||
NM_148957.3 | Intron | NP_683760.1 | ||||
XM_005266445.2 | Intron | XP_005266502.1 | ||||
XM_005266446.1 | Intron | XP_005266503.1 | ||||
XM_011535146.1 | Intron | XP_011533448.1 | ||||
XM_017020651.1 | Intron | XP_016876140.1 | ||||
XM_017020652.1 | Intron | XP_016876141.1 |
Set Membership: |
HapMap |