Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604209 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPNE5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPNE5 - copine 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001314017.1 | 1015 | Intron | NP_001300946.1 | |||
NM_001314018.1 | 1015 | Missense Mutation | CAC,CGC | H,R 290 | NP_001300947.1 | |
NM_001314019.1 | 1015 | Missense Mutation | CAC,CGC | H,R 232 | NP_001300948.1 | |
NM_001314020.1 | 1015 | Missense Mutation | CAC,CGC | H,R 232 | NP_001300949.1 | |
NM_020939.1 | 1015 | Missense Mutation | CAC,CGC | H,R 582 | NP_065990.1 | |
XM_005249247.2 | 1015 | Missense Mutation | CAC,CGC | H,R 599 | XP_005249304.1 | |
XM_011514768.1 | 1015 | Intron | XP_011513070.1 | |||
XM_011514769.1 | 1015 | Intron | XP_011513071.1 | |||
XM_011514770.1 | 1015 | Intron | XP_011513072.1 | |||
XM_011514771.2 | 1015 | Intron | XP_011513073.1 | |||
XM_011514772.1 | 1015 | Intron | XP_011513074.1 | |||
XM_011514773.2 | 1015 | Intron | XP_011513075.1 | |||
XM_011514775.2 | 1015 | Intron | XP_011513077.1 | |||
XM_017011139.1 | 1015 | Intron | XP_016866628.1 |
RAB44 - RAB44, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |