Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600864 MIM: 603877 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSNK1D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSNK1D - casein kinase 1 delta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6787 - microRNA 6787 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC16A3 - solute carrier family 16 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042422.2 | 1678 | Intron | NP_001035887.1 | |||
NM_001042423.2 | 1678 | Intron | NP_001035888.1 | |||
NM_001206950.1 | 1678 | Intron | NP_001193879.1 | |||
NM_001206951.1 | 1678 | Intron | NP_001193880.1 | |||
NM_001206952.1 | 1678 | Intron | NP_001193881.1 | |||
NM_004207.3 | 1678 | Intron | NP_004198.1 | |||
XM_011523607.1 | 1678 | UTR 5 | XP_011521909.1 |