Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGTGGTACTGAGGCAGACGTTGT[A/G]GCGGAGAAGGGCAGTGGCTGTAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602694 | ||||||||||||||||||||
Literature Links: |
NDUFS4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NDUFS4 - NADH:ubiquinone oxidoreductase subunit S4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318051.1 | 74 | Nonsense Mutation | TAG,TGG | *,W 15 | NP_001304980.1 | |
NM_002495.3 | 74 | Nonsense Mutation | TAG,TGG | *,W 15 | NP_002486.1 | |
XM_017009491.1 | 74 | Nonsense Mutation | TAG,TGG | *,W 15 | XP_016864980.1 |