Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCCGGGCGGCCGCGGCCCGTGC[C/T]GCGCTCGGTGAACTCGCGCGAGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608537 | ||||||||||||||||||||
Literature Links: |
VHL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VHL - von Hippel-Lindau tumor suppressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000551.3 | 401 | Missense Mutation | CCG,CTG | P,L 63 | NP_000542.1 | |
NM_198156.2 | 401 | Missense Mutation | CCG,CTG | P,L 63 | NP_937799.1 |