Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCTACAATGAAATTGCAGGTCC[A/G]ATTGCTGAATGCAGCTATTGCTAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138160 | ||||||||||||||||||||
Literature Links: |
SLC2A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC2A2 - solute carrier family 2 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000340.1 | 1639 | Missense Mutation | CGG,TGG | R,W 444 | NP_000331.1 | |
NM_001278658.1 | 1639 | Missense Mutation | CGG,TGG | R,W 325 | NP_001265587.1 | |
NM_001278659.1 | 1639 | Missense Mutation | CGG,TGG | R,W 271 | NP_001265588.1 |