Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCTGGCTGCAACAGCGTTTGCT[A/G]TGACCAATTCTTCCCCATCTCCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 304040 | ||||||||||||||||||||
Literature Links: |
GJB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GJB1 - gap junction protein beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000166.5 | 340 | Missense Mutation | TAT,TGT | Y,C 65 | NP_000157.1 | |
NM_001097642.2 | 340 | Missense Mutation | TAT,TGT | Y,C 65 | NP_001091111.1 | |
XM_011530907.2 | 340 | Missense Mutation | TAT,TGT | Y,C 65 | XP_011529209.1 | |
XM_017029408.1 | 340 | Missense Mutation | TAT,TGT | Y,C 65 | XP_016884897.1 |