Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147569 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFNGR2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IFNGR2 - interferon gamma receptor 2 (interferon gamma transducer 1) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005534.3 | 821 | Missense Mutation | ACG,AGG | T,R 58 | NP_005525.2 | |
XM_005260969.2 | 821 | Missense Mutation | ACG,AGG | T,R 77 | XP_005261026.1 | |
XM_011529553.1 | 821 | Missense Mutation | ACG,AGG | T,R 83 | XP_011527855.1 | |
XM_011529554.2 | 821 | Missense Mutation | ACG,AGG | T,R 60 | XP_011527856.1 |
Set Membership: |
HapMap |