Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCGGCCGGGCTCCCTGGAGAGG[A/G]AGTGCAAGGAGGAGCAGTGCTCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613878 | ||||||||||||||||||||
Literature Links: |
F7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
F7 - coagulation factor VII | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000131.4 | 214 | Missense Mutation | AAG,GAG | K,E 76 | NP_000122.1 | |
NM_001267554.1 | 214 | Intron | NP_001254483.1 | |||
NM_019616.3 | 214 | Missense Mutation | AAG,GAG | K,E 54 | NP_062562.1 |