Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGAAAAATCAGCATTTTCAGAA[A/T]AGGCATGCTGAATGCCCATCTTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 314200 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERPINA7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SERPINA7 - serpin family A member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000354.5 | 1031 | Missense Mutation | TAT,TTT | Y,F 329 | NP_000345.2 | |
XM_005262180.3 | 1031 | Missense Mutation | TAT,TTT | Y,F 329 | XP_005262237.1 | |
XM_006724683.2 | 1031 | Missense Mutation | TAT,TTT | Y,F 329 | XP_006724746.1 |