Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGCTGAGGTGAGTCCTGGAGGT[C/G]CGTGGTTGGGGCGGAGAGCATTATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604737 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GDAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GDAP2 - ganglioside induced differentiation associated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135589.2 | 245 | Intron | NP_001129061.1 | |||
NM_017686.3 | 245 | Intron | NP_060156.1 | |||
XM_005270959.2 | 245 | Intron | XP_005271016.1 | |||
XM_011541621.2 | 245 | Intron | XP_011539923.1 | |||
XM_011541622.1 | 245 | Intron | XP_011539924.1 | |||
XM_011541623.2 | 245 | Intron | XP_011539925.1 | |||
XM_011541624.2 | 245 | UTR 5 | XP_011539926.1 | |||
XM_017001531.1 | 245 | Intron | XP_016857020.1 | |||
XM_017001532.1 | 245 | UTR 5 | XP_016857021.1 |
WDR3 - WD repeat domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006784.2 | 245 | Intron | NP_006775.1 |