Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTAACAAAAATTTGTGTTTTGTT[C/T]ATTAACTTTTTTTAATAACCCCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
EGFLAM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EGFLAM - EGF like, fibronectin type III and laminin G domains | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205301.1 | Intron | NP_001192230.1 | ||||
NM_152403.3 | Intron | NP_689616.2 | ||||
NM_182798.2 | Intron | NP_877950.1 | ||||
NM_182801.2 | Intron | NP_877953.1 |
EGFLAM-AS2 - EGFLAM antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |