Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTTCTCGCCGGCTTCCCGAGTTC[C/T]CGGGGGCGGGGCTTGTGTTTTTACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 305900 MIM: 300248 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
G6PD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
G6PD - glucose-6-phosphate dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000402.4 | 224 | Intron | NP_000393.4 | |||
NM_001042351.2 | 224 | UTR 5 | NP_001035810.1 |
IKBKG - inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099856.4 | 224 | Intron | NP_001093326.2 | |||
NM_001099857.2 | 224 | UTR 5 | NP_001093327.1 | |||
NM_001145255.2 | 224 | UTR 5 | NP_001138727.1 | |||
NM_001321396.1 | 224 | Intron | NP_001308325.1 | |||
NM_001321397.1 | 224 | UTR 5 | NP_001308326.1 | |||
NM_003639.4 | 224 | Intron | NP_003630.1 |