Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCACAGACAGACAACCCCTCCTT[C/T]CCACCCCTGCCCTGTCCCCCATCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614297 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C19orf12 - chromosome 19 open reading frame 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031726.3 | 2919 | UTR 3 | NP_001026896.2 | |||
NM_001256046.1 | 2919 | UTR 3 | NP_001242975.1 | |||
NM_001256047.1 | 2919 | UTR 3 | NP_001242976.1 | |||
NM_001282929.1 | 2919 | UTR 3 | NP_001269858.1 | |||
NM_001282930.1 | 2919 | UTR 3 | NP_001269859.1 | |||
NM_001282931.1 | 2919 | UTR 3 | NP_001269860.1 | |||
NM_031448.4 | 2919 | UTR 3 | NP_113636.2 |