Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATGGCGGCTCTCAGGCTGGCGC[G/T]CTCCGTGCTGCTGGGGCTTTGAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606576 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TAF3 - TATA-box binding protein associated factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031923.3 | 48 | UTR 5 | NP_114129.1 | |||
XM_011519741.1 | 48 | Missense Mutation | GCT,TCT | A,S 25 | XP_011518043.1 |