Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTTTAGTGCAGCGAGGAAGTTT[G/T]CGCTTCTGTACATGTGTTTGTGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 139150 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RASA1 - RAS p21 protein activator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002890.2 | 353 | Intron | NP_002881.1 | |||
NM_022650.2 | 353 | Intron | NP_072179.1 | |||
XM_011543525.1 | 353 | UTR 5 | XP_011541827.1 | |||
XM_011543527.2 | 353 | UTR 5 | XP_011541829.1 |
Set Membership: |
HapMap |