Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTTAACAAAAAAGCCCCACGCCC[C/G]CTTTGACGGCTTAGAGCCCACCTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608927 | ||||||||||||||||||||
Literature Links: |
COL26A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COL26A1 - collagen type XXVI alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278563.2 | 445 | Intron | NP_001265492.1 | |||
NM_133457.4 | 445 | Intron | NP_597714.2 | |||
XM_017011743.1 | 445 | UTR 5 | XP_016867232.1 | |||
XM_017011744.1 | 445 | Intron | XP_016867233.1 | |||
XM_017011745.1 | 445 | Intron | XP_016867234.1 |