Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGCCCGACCAATGTGGAAGCCT[C/T]CTCGGGTGCTCCGCCCTCCCCGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 248611 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCKDHB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCKDHB - branched chain keto acid dehydrogenase E1 subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000056.4 | 14 | UTR 5 | NP_000047.1 | |||
NM_001318975.1 | 14 | UTR 5 | NP_001305904.1 | |||
NM_183050.3 | 14 | UTR 5 | NP_898871.1 | |||
XM_005248756.4 | 14 | Intron | XP_005248813.1 | |||
XM_011536023.2 | 14 | Intron | XP_011534325.1 | |||
XM_011536024.2 | 14 | Intron | XP_011534326.1 | |||
XM_011536025.2 | 14 | Intron | XP_011534327.1 | |||
XM_011536026.2 | 14 | Intron | XP_011534328.1 |