Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTCTAATCGGCCCATCTCTTTCC[C/G]TTTCTGTGTGGTTACCAGGTTACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611485 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP4F12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP4F12 - cytochrome P450 family 4 subfamily F member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023944.3 | Intron | NP_076433.3 | ||||
XM_006722850.3 | Intron | XP_006722913.1 | ||||
XM_011528202.2 | Intron | XP_011526504.1 | ||||
XM_011528203.2 | Intron | XP_011526505.1 | ||||
XM_011528204.2 | Intron | XP_011526506.1 | ||||
XM_011528205.2 | Intron | XP_011526507.1 | ||||
XM_011528207.2 | Intron | XP_011526509.1 | ||||
XM_011528208.2 | Intron | XP_011526510.1 | ||||
XM_017027172.1 | Intron | XP_016882661.1 |