Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGATGCCAACCTTTCTCCCTCACA[A/G]AGACAACCCTTTATCCTGGGAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610590 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP23 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP23 - Rho GTPase activating protein 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199417.1 | Intron | NP_001186346.1 | ||||
XM_006721991.1 | Intron | XP_006722054.1 | ||||
XM_006721992.3 | Intron | XP_006722055.1 | ||||
XM_006721993.3 | Intron | XP_006722056.1 | ||||
XM_006721994.3 | Intron | XP_006722057.1 | ||||
XM_011525073.1 | Intron | XP_011523375.1 | ||||
XM_011525074.2 | Intron | XP_011523376.1 | ||||
XM_011525075.2 | Intron | XP_011523377.1 | ||||
XM_011525076.2 | Intron | XP_011523378.1 | ||||
XM_011525077.2 | Intron | XP_011523379.1 | ||||
XM_017024902.1 | Intron | XP_016880391.1 |