Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCGTGTAGCGCAGCTGCTTCTG[A/G]TCCGGGAGCTCGGCAGGGCTGGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606839 MIM: 605047 MIM: 611780 | ||||||||||||||||||||
Literature Links: |
CDHR5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDHR5 - cadherin related family member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IRF7 - interferon regulatory factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001572.3 | 1466 | Silent Mutation | GAC,GAT | D,D 339 | NP_001563.2 | |
NM_004029.2 | 1466 | Silent Mutation | GAC,GAT | D,D 310 | NP_004020.1 | |
NM_004031.2 | 1466 | Silent Mutation | GAC,GAT | D,D 352 | NP_004022.2 | |
XM_005252906.3 | 1466 | Silent Mutation | GAC,GAT | D,D 352 | XP_005252963.1 | |
XM_005252907.3 | 1466 | Silent Mutation | GAC,GAT | D,D 351 | XP_005252964.1 | |
XM_005252909.3 | 1466 | Silent Mutation | GAC,GAT | D,D 323 | XP_005252966.1 | |
XM_011520066.2 | 1466 | Silent Mutation | GAC,GAT | D,D 338 | XP_011518368.1 | |
XM_017017674.1 | 1466 | Silent Mutation | GAC,GAT | D,D 46 | XP_016873163.1 |
PHRF1 - PHD and ring finger domains 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |