Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGAAACGGAGGCAAGAAAGGGGG[C/G]TACCTCAGGAGCGAGGGACAAAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615514 | ||||||||||||||||||||
Literature Links: |
CDK12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDK12 - cyclin dependent kinase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015083.2 | 70 | UTR 5 | NP_055898.1 | |||
NM_016507.3 | 70 | UTR 5 | NP_057591.2 |