Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCAAATATGGAAGTGTGCAAGT[C/G]TTGAACACATTAGTTTATACAATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615882 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RABGAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RABGAP1 - RAB GTPase activating protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012197.3 | 52 | Intron | NP_036329.3 | |||
XM_011518440.2 | 52 | Intron | XP_011516742.1 | |||
XM_011518441.2 | 52 | UTR 5 | XP_011516743.1 | |||
XM_011518442.2 | 52 | Intron | XP_011516744.1 | |||
XM_011518444.2 | 52 | Intron | XP_011516746.1 | |||
XM_017014567.1 | 52 | Intron | XP_016870056.1 | |||
XM_017014568.1 | 52 | UTR 5 | XP_016870057.1 | |||
XM_017014569.1 | 52 | Intron | XP_016870058.1 | |||
XM_017014570.1 | 52 | Intron | XP_016870059.1 |