Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGAAGGAATTCTAGCAAATGAAAC[A/T]AATAGGGACACCTTTTTTCAGAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RALGPS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RALGPS2 - Ral GEF with PH domain and SH3 binding motif 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286247.1 | Intron | NP_001273176.1 | ||||
NM_152663.4 | Intron | NP_689876.2 | ||||
XM_006711410.3 | Intron | XP_006711473.1 | ||||
XM_006711411.3 | Intron | XP_006711474.1 | ||||
XM_011509688.1 | Intron | XP_011507990.1 | ||||
XM_017001591.1 | Intron | XP_016857080.1 |