Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGGTGCGTTTTACAATAAGTGGA[A/G]AAAGGAAGGGCGTGTGGACAGGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605904 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDLIM5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDLIM5 - PDZ and LIM domain 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011513.3 | Intron | NP_001011513.3 | ||||
NM_001011515.2 | Intron | NP_001011515.1 | ||||
NM_001011516.2 | Intron | NP_001011516.1 | ||||
NM_001256425.1 | Intron | NP_001243354.1 | ||||
NM_001256426.1 | Intron | NP_001243355.1 | ||||
NM_001256427.1 | Intron | NP_001243356.1 | ||||
NM_001256428.1 | Intron | NP_001243357.1 | ||||
NM_001256429.1 | Intron | NP_001243358.1 | ||||
NM_006457.4 | Intron | NP_006448.4 | ||||
XM_005262693.4 | Intron | XP_005262750.1 | ||||
XM_005262695.4 | Intron | XP_005262752.1 | ||||
XM_005262696.4 | Intron | XP_005262753.1 | ||||
XM_005262698.4 | Intron | XP_005262755.1 | ||||
XM_006714066.3 | Intron | XP_006714129.1 | ||||
XM_006714068.3 | Intron | XP_006714131.1 | ||||
XM_006714069.3 | Intron | XP_006714132.1 | ||||
XM_006714070.3 | Intron | XP_006714133.1 | ||||
XM_011531543.2 | Intron | XP_011529845.1 | ||||
XM_017007657.1 | Intron | XP_016863146.1 | ||||
XM_017007658.1 | Intron | XP_016863147.1 |