Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTTAGCTCTTTGTGCTCCAGAGA[C/G]ACATTTTACAGTCATGATTTGGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603110 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMAD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMAD5 - SMAD family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001419.2 | Intron | NP_001001419.1 | ||||
NM_001001420.2 | Intron | NP_001001420.1 | ||||
NM_005903.6 | Intron | NP_005894.3 | ||||
XM_017009470.1 | Intron | XP_016864959.1 |
SMAD5-AS1 - SMAD5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |