Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMDHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AMDHD1 - amidohydrolase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152435.2 | 113 | Missense Mutation | AGC,GGC | S,G 3 | NP_689648.2 |
CCDC38 - coiled-coil domain containing 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182496.2 | 113 | Intron | NP_872302.2 | |||
XM_006719229.2 | 113 | Intron | XP_006719292.1 | |||
XM_011537883.2 | 113 | Intron | XP_011536185.1 | |||
XM_011537884.2 | 113 | Intron | XP_011536186.1 | |||
XM_011537886.2 | 113 | Intron | XP_011536188.1 | |||
XM_011537887.2 | 113 | Intron | XP_011536189.1 | |||
XM_011537888.2 | 113 | Intron | XP_011536190.1 | |||
XM_011537889.1 | 113 | Intron | XP_011536191.1 |
Set Membership: |
HapMap |