Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAAGACGTTATTTCACAGAGCTA[C/T]GTGGCAACACACAGAAACACTGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609768 MIM: 604532 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLOC1S2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BLOC1S2 - biogenesis of lysosomal organelles complex 1 subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PKD2L1 - polycystin 2 like 1, transient receptor potential cation channel | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253837.1 | 2605 | Intron | NP_001240766.1 | |||
NM_016112.2 | 2605 | Intron | NP_057196.2 | |||
XM_011540323.2 | 2605 | UTR 3 | XP_011538625.1 | |||
XM_011540325.2 | 2605 | UTR 3 | XP_011538627.1 | |||
XM_017016875.1 | 2605 | UTR 3 | XP_016872364.1 | |||
XM_017016876.1 | 2605 | UTR 3 | XP_016872365.1 |
Set Membership: |
HapMap |