Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGAGAAGCACCAGCAGGCAGTAA[C/G]AGCCAACCCTTAGCCATTGCTAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616805 MIM: 192320 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYCT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYCT1 - myc target 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VIP - vasoactive intestinal peptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003381.3 | 2936 | UTR 5 | NP_003372.1 | |||
NM_194435.2 | 2936 | UTR 5 | NP_919416.1 | |||
XM_005267135.3 | 2936 | Intron | XP_005267192.1 | |||
XM_006715562.3 | 2936 | UTR 5 | XP_006715625.1 |