Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTCTCCTTGGGATGATGGCTGG[C/T]TAGTCAGCCTTGCATGTATTCCTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601597 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTG2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BTG2 - BTG anti-proliferation factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006763.2 | 1725 | UTR 3 | NP_006754.1 |
LINC01136 - long intergenic non-protein coding RNA 1136 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |