Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATTTGATAATACCAAAAATTAC[A/C]TACTTATATACATAATTGTGGAGAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608959 | |||||||||||||||||||||||
Literature Links: |
DPH3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
DPH3 - diphthamide biosynthesis 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OXNAD1 - oxidoreductase NAD binding domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138381.3 | Intron | NP_612390.1 | ||||
XM_005265559.3 | Intron | XP_005265616.1 | ||||
XM_006713405.1 | Intron | XP_006713468.1 | ||||
XM_006713406.2 | Intron | XP_006713469.1 | ||||
XM_011534230.2 | Intron | XP_011532532.1 | ||||
XM_011534232.2 | Intron | XP_011532534.1 | ||||
XM_011534233.2 | Intron | XP_011532535.1 | ||||
XM_011534234.2 | Intron | XP_011532536.1 | ||||
XM_011534235.2 | Intron | XP_011532537.1 | ||||
XM_017007485.1 | Intron | XP_016862974.1 | ||||
XM_017007486.1 | Intron | XP_016862975.1 | ||||
XM_017007487.1 | Intron | XP_016862976.1 | ||||
XM_017007488.1 | Intron | XP_016862977.1 | ||||
XM_017007489.1 | Intron | XP_016862978.1 | ||||
XM_017007490.1 | Intron | XP_016862979.1 | ||||
XM_017007491.1 | Intron | XP_016862980.1 | ||||
XM_017007492.1 | Intron | XP_016862981.1 | ||||
XM_017007493.1 | Intron | XP_016862982.1 | ||||
XM_017007494.1 | Intron | XP_016862983.1 | ||||
XM_017007495.1 | Intron | XP_016862984.1 | ||||
XM_017007496.1 | Intron | XP_016862985.1 |