Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGGTCTGGGCCAGGGAGGCGAT[C/T]CCCTCCGATGCGCGGGACAGAGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605467 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF274 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF274 - zinc finger protein 274 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278734.1 | 206 | Intron | NP_001265663.1 | |||
NM_016324.3 | 206 | Intron | NP_057408.2 | |||
NM_016325.3 | 206 | Intron | NP_057409.1 | |||
NM_133502.2 | 206 | Intron | NP_598009.1 | |||
XM_011526327.1 | 206 | Intron | XP_011524629.1 | |||
XM_011526328.2 | 206 | Intron | XP_011524630.1 | |||
XM_017026174.1 | 206 | Intron | XP_016881663.1 | |||
XM_017026175.1 | 206 | UTR 5 | XP_016881664.1 |