Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGGATTTTTGCTCTTTCTAAGT[A/C]ATGCATATTTATATTATCTTCAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608533 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO38 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO38 - F-box protein 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271723.1 | Intron | NP_001258652.1 | ||||
NM_030793.4 | Intron | NP_110420.3 | ||||
XM_005268513.1 | Intron | XP_005268570.1 | ||||
XM_006714797.1 | Intron | XP_006714860.1 | ||||
XM_011537684.2 | Intron | XP_011535986.1 | ||||
XM_017009899.1 | Intron | XP_016865388.1 | ||||
XM_017009900.1 | Intron | XP_016865389.1 | ||||
XM_017009901.1 | Intron | XP_016865390.1 | ||||
XM_017009902.1 | Intron | XP_016865391.1 |