Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCCGTCCAGCCTGGTTGTTGAAG[C/T]GTCCCCGTGGTCCCCCGGGGTCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600414 | ||||||||||||||||||||
Literature Links: |
PEX5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PEX5 - peroxisomal biogenesis factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000319.4 | 364 | Intron | NP_000310.2 | |||
NM_001131023.1 | 364 | Intron | NP_001124495.1 | |||
NM_001131024.1 | 364 | Intron | NP_001124496.1 | |||
NM_001131025.1 | 364 | UTR 5 | NP_001124497.1 | |||
NM_001131026.1 | 364 | UTR 5 | NP_001124498.1 | |||
NM_001300789.1 | 364 | Intron | NP_001287718.1 |