Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCGGGCTGCCATCTTGCTCTAAG[C/T]GAAAGTGAAAGAAAAGTCGGCAGCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186711 MIM: 607081 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD27 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CD27 - CD27 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242.4 | 75 | Intron | NP_001233.1 | |||
XM_011521042.2 | 75 | Intron | XP_011519344.1 | |||
XM_017020232.1 | 75 | Intron | XP_016875721.1 | |||
XM_017020233.1 | 75 | Intron | XP_016875722.1 | |||
XM_017020234.1 | 75 | Intron | XP_016875723.1 |
CD27-AS1 - CD27 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAPBPL - TAP binding protein like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018009.4 | 75 | UTR 5 | NP_060479.3 | |||
XM_005253700.3 | 75 | UTR 5 | XP_005253757.1 | |||
XM_017019545.1 | 75 | Intron | XP_016875034.1 | |||
XM_017019546.1 | 75 | Intron | XP_016875035.1 |
Set Membership: |
HapMap |