Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGACTACCCGGGACTGCGGCCGCC[G/T]CGTCAGGTGCAGCGCCAGGAGCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603746 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLIT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLIT2 - slit guidance ligand 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289135.2 | 539 | UTR 5 | NP_001276064.1 | |||
NM_001289136.2 | 539 | UTR 5 | NP_001276065.1 | |||
NM_004787.3 | 539 | UTR 5 | NP_004778.1 | |||
XM_005248211.3 | 539 | UTR 5 | XP_005248268.1 | |||
XM_006713986.3 | 539 | UTR 5 | XP_006714049.1 | |||
XM_011513909.2 | 539 | Intron | XP_011512211.1 | |||
XM_011513910.2 | 539 | Intron | XP_011512212.2 | |||
XM_017008845.1 | 539 | Intron | XP_016864334.1 |