Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGACCGTAGTTTCTTGATGGCTAA[C/G]ATGTTTGGAGGGTGGCTCATGTATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616865 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAPOLG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAPOLG - poly(A) polymerase gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022894.3 | Intron | NP_075045.2 | ||||
XM_005264500.3 | Intron | XP_005264557.1 | ||||
XM_005264501.1 | Intron | XP_005264558.1 | ||||
XM_011533041.1 | Intron | XP_011531343.1 | ||||
XM_017004723.1 | Intron | XP_016860212.1 | ||||
XM_017004724.1 | Intron | XP_016860213.1 |