Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTTTTTTTACAAGTAAGCGTC[C/T]ATGCAGGCATCACAAACTTTGGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 167414 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAX5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAX5 - paired box 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001280547.1 | 8097 | UTR 3 | NP_001267476.1 | |||
NM_001280548.1 | 8097 | UTR 3 | NP_001267477.1 | |||
NM_001280549.1 | 8097 | UTR 3 | NP_001267478.1 | |||
NM_001280550.1 | 8097 | UTR 3 | NP_001267479.1 | |||
NM_001280551.1 | 8097 | UTR 3 | NP_001267480.1 | |||
NM_001280552.1 | 8097 | UTR 3 | NP_001267481.1 | |||
NM_001280553.1 | 8097 | UTR 3 | NP_001267482.1 | |||
NM_001280554.1 | 8097 | UTR 3 | NP_001267483.1 | |||
NM_001280555.1 | 8097 | UTR 3 | NP_001267484.1 | |||
NM_001280556.1 | 8097 | UTR 3 | NP_001267485.1 | |||
NM_016734.2 | 8097 | UTR 3 | NP_057953.1 | |||
XM_005251481.4 | 8097 | UTR 3 | XP_005251538.1 | |||
XM_011517896.2 | 8097 | UTR 3 | XP_011516198.1 | |||
XM_011517897.2 | 8097 | UTR 3 | XP_011516199.1 |