Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGCGCCGCTGTGATTGGTGAGGC[C/T]CCGCCAGGGGCGGAGACGACCTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602638 MIM: 600899 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCM4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MCM4 - minichromosome maintenance complex component 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005914.3 | 690 | UTR 5 | NP_005905.2 | |||
NM_182746.2 | 690 | UTR 5 | NP_877423.1 |
PRKDC - protein kinase, DNA-activated, catalytic polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081640.1 | 690 | Intron | NP_001075109.1 | |||
NM_006904.6 | 690 | Intron | NP_008835.5 |