Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCCCACTCCATGACCTCTGGAC[A/G]CCCAGTCGGGTCCAGGCGCGATAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616923 MIM: 180474 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102724450 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC102724450 - uncharacterized LOC102724450 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF207 - ring finger protein 207 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207396.2 | Intron | NP_997279.2 | ||||
XM_011541439.2 | Intron | XP_011539741.1 | ||||
XM_017001259.1 | Intron | XP_016856748.1 |
RPL22 - ribosomal protein L22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |