Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACTCGGGCGAAACGGCGTGTGTT[A/T]TGCCTAAGACTCTGCCAGTGTTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSP90B2P PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSP90B2P - heat shock protein 90 beta family member 2, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC28 - leucine rich repeat containing 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284400.2 | 36 | Intron | NP_001271329.1 | |||
NM_001321675.1 | 36 | UTR 5 | NP_001308604.1 | |||
NM_001321676.1 | 36 | Intron | NP_001308605.1 | |||
NM_001321677.1 | 36 | Intron | NP_001308606.1 | |||
NM_001321678.1 | 36 | Intron | NP_001308607.1 | |||
NM_001321679.1 | 36 | Intron | NP_001308608.1 | |||
NM_001321680.1 | 36 | UTR 5 | NP_001308609.1 | |||
NM_144598.4 | 36 | Intron | NP_653199.2 | |||
XM_006720389.3 | 36 | Intron | XP_006720452.1 | |||
XM_011521218.2 | 36 | Intron | XP_011519520.1 | |||
XM_011521220.1 | 36 | Intron | XP_011519522.1 | |||
XM_011521221.2 | 36 | Intron | XP_011519523.1 | |||
XM_017021914.1 | 36 | UTR 5 | XP_016877403.1 |