Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCACGTGTGACATTCTGTATCC[A/G]GTCTATCGGTTATCCCAAGGAGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613734 MIM: 612981 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC115 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCDC115 - coiled-coil domain containing 115 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321118.1 | 491 | Intron | NP_001308047.1 | |||
NM_001321119.1 | 491 | Intron | NP_001308048.1 | |||
NM_032357.3 | 491 | Intron | NP_115733.2 |
IMP4 - IMP4 homolog, U3 small nucleolar ribonucleoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320304.1 | 491 | UTR 5 | NP_001307233.1 | |||
NM_001320305.1 | 491 | UTR 5 | NP_001307234.1 | |||
NM_001320306.1 | 491 | UTR 5 | NP_001307235.1 | |||
NM_001320307.1 | 491 | UTR 5 | NP_001307236.1 | |||
NM_001320309.1 | 491 | UTR 5 | NP_001307238.1 | |||
NM_001320310.1 | 491 | UTR 5 | NP_001307239.1 | |||
NM_001320311.1 | 491 | UTR 5 | NP_001307240.1 | |||
NM_033416.2 | 491 | UTR 5 | NP_219484.1 |
Set Membership: |
HapMap |