Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGCCTGGGCGCGGGAACGCATC[C/T]GCACTGCGAAAGAGATGCCAGGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602294 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOXA1 - forkhead box A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC6 - tetratricopeptide repeat domain 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001310135.1 | Intron | NP_001297064.1 | ||||
XM_011537430.2 | Intron | XP_011535732.1 | ||||
XM_011537431.2 | Intron | XP_011535733.1 | ||||
XM_011537432.2 | Intron | XP_011535734.1 | ||||
XM_017021254.1 | Intron | XP_016876743.1 | ||||
XM_017021255.1 | Intron | XP_016876744.1 | ||||
XM_017021256.1 | Intron | XP_016876745.1 | ||||
XM_017021257.1 | Intron | XP_016876746.1 |